Diethylene glycol monoethyl ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-90-0
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00002872
:134.18 g/mol
Merck Index:13,01809
Boiling Pt:197 °C (1013 hPa)
Melting Pt:–80 °C
Density:0,99 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:93 °C
Storage Temperature:Ambient
EINECS:203-919-7
REACH:01-2119475105-42
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-78 °C
- Molar Mass:134.17 g/mol
- Boiling Point:202 °C (1013 hPa)
- Vapor Pressure:0.13 hPa (20 °C)
- Flash Point:96 °C
- Refractive Index:1.4293 (20 °C, 589 nm)
- Explosion Limit:1.8 - 12.2 %(V)
- Density:0.99 g/cm3 (20 °C)
- Ignition Point:204 °C
- Evaporation number:1200
Specification Test Results
Related Information
Frequently Bought Together