Diethylene glycol monoethyl ether 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-90-0
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00002872
:134.18 g/mol
Merck Index:13,01809
Boiling Pt:197 °C (760 mmHg)
Melting Pt:–80 °C
Density:0,99 g/cm³
Flash Pt:94 °C
Storage Temperature:Ambient
EINECS:203-919-7
REACH:01-2119475105-42
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4250 to 1.4280 (20°C, 589 nm)
- GC:≥98.0 %
- Sulfate (SO4):≤0.003 %
- Potassium carbonate:≤2.0 % (K2CO3)
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.002 %
- Nickel (Ni):≤0.001 %
- Sodium (Na):≤0.05 %
- Heavy metals (ICP-OES):≤0.001 %
- Nitrogen compounds (as N):≤0.001 %
Specification Test Results
Frequently Bought Together