


3-Diethylaminopropylamine 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-78-9
:
Formula:C₇H₁₈N₂
MDL Number:MFCD00008218
:130.23 g/mol
Boiling Pt:168…171 °C (760 mmHg)
Melting Pt:–60 °C
Density:0,82 g/cm³
Flash Pt:53 °C
Storage Temperature:Ambient
EINECS:203-236-4
UN:2684
ADR:3,III
:GHS07
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4406 to 1.4426 (20°C, 589 nm)
- Water:≤0.3 %
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together