Anti-p21 Waf1/Cip1 Rabbit Monoclonal Antibody [clone: 12D1] (Alexa Fluor 488)
CSTA5487S
- Antibody type:Primary
- Antigen symbol:p21 Waf1/Cip1
- Clonality:Monoclonal
- Clone:12D1
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Sequence:CPRSGEQAEGSPGGPGDSQGRKRRQT
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: p21 Waf1/Cip1
Clonality: Monoclonal
Clone: 12D1
Conjugation: Alexa Fluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey