Anti-Trail-R3/DcR1 Rabbit Polyclonal Antibody
Catalog # CSTA4756S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Trail-R3/DcR1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CGTPAPAAEETMTTSPGTPASSHYLS
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Trail-R3/DcR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat