Order Entry
Ireland
ContactUsLinkComponent
Anti-Pim-1 Rabbit Monoclonal Antibody [clone: C93F2]
Anti-Pim-1 Rabbit Monoclonal Antibody [clone: C93F2]
Catalog # CSTA3247S
Supplier:  Cell Signaling
Anti-Pim-1 Rabbit Monoclonal Antibody [clone: C93F2]
Catalog # CSTA3247S
Supplier:  Cell Signaling

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Pim-1
  • Clonality:
    Monoclonal
  • Clone:
    C93F2
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Sequence:
    CKHVEKDRISDWGELPNGTR
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    Ambient
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Pim-1
Clonality: Monoclonal
Clone: C93F2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse