Anti-Torc3 Rabbit Polyclonal Antibody
CSTA2768S
- Antibody type:Primary
- Antigen symbol:Torc3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Sequence:CSSQPLDESWPRQQPPWKDEKHPG
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Torc3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat