Anti-XRCC1 Rabbit Polyclonal Antibody
CSTA2735S
- Antibody type:Primary
- Antigen symbol:XRCC1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Sequence:GKPRGEGTEPRRPRAGPC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: XRCC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human