Anti-TMP21 Rabbit Polyclonal Antibody
CSTA2076S
- Antibody type:Primary
- Antigen symbol:TMP21
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Sequence:CAGHILYSKEDATKGKFAFTTEDYD
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: TMP21
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Monkey