Anti-MYH Rabbit Monoclonal Antibody [clone: D13D4]
Catalog # CSTA6248S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:MYH
- Clonality:Monoclonal
- Clone:D13D4
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CGVVNFPRKASRKPPREESSAT
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MYH
Clonality: Monoclonal
Clone: D13D4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human