Anti-ARPP-19 (Ser62) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA5240S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:ARPP-19 (Ser62) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Antigen synonyms:ENSA (Ser67) phosphate
- Sequence:CGQKYFDsGDYN
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ARPP-19 (Ser62) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat