Anti-acetyl-CoA carboxylase 1 (Ser80) phosphate Rabbit Polyclonal Antibody
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: acetyl-CoA carboxylase 1 (Ser80) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat;Monkey
Specifications
- Catalog No:
- CSTA3661L
- CSTA3661S
- Antigen symbol:
- acetyl-CoA carboxylase 1 (Ser80) phosphate
- acetyl-CoA carboxylase 1 (Ser80) phosphate
- Antigen synonyms:
- acetyl-CoA carboxylase 2 (Ser222) phosphate
- acetyl-CoA carboxylase 2 (Ser222) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CHMRSSMsGLHLVK
- CHMRSSMsGLHLVK
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together