Anti-TORC1 Rabbit Monoclonal Antibody [clone: C71D11]
Catalog # CSTA2587S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:TORC1
- Clonality:Monoclonal
- Clone:C71D11
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Sequence:CANQSPTSPVSNQGFSPGSSPQHT
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TORC1
Clonality: Monoclonal
Clone: C71D11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat