Anti-AMPK-gamma3 Rabbit Polyclonal Antibody
Catalog # CSTA2550S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:AMPK-gamma3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Sequence:CTFPKTTPLAQADPAGVGTPPTGWD
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: AMPK-gamma3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human