Anti-Mitofusin-2 Rabbit Monoclonal Antibody [clone: D1E9]
Catalog # CSTA11925S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Mitofusin-2
- Clonality:Monoclonal
- Clone:D1E9
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster,Monkey,Human
- Western blot:Yes
- Sequence:CRALMGYNDQVQRPIPLTPANPS
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Mitofusin-2
Clonality: Monoclonal
Clone: D1E9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Hamster;Monkey