Anti-HIP Mouse Monoclonal Antibody [clone: 10B7-C3]
CSTA8014S
- Antibody type:Primary
- Antigen symbol:HIP
- Clonality:Monoclonal
- Clone:10B7-C3
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:Purified recombinant Rat HIP (full-length) expressed in E. Coli.
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: HIP
Clonality: Monoclonal
Clone: 10B7-C3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Monkey