Anti-JMJD2A Rabbit Monoclonal Antibody [clone: C37E5]
Catalog # CSTA5328S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:JMJD2A
- Clonality:Monoclonal
- Clone:C37E5
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CRPTHSSVRQVEDSLPFPDYSD
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: JMJD2A
Clonality: Monoclonal
Clone: C37E5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human