Anti-Grb2 Rabbit Polyclonal Antibody
Catalog # CSTA3972S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Grb2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Hamster,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Sequence:CFPRNyVTPVNRNV
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Grb2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Hamster, Monkey, Bovine