Anti-GFAP Mouse Monoclonal Antibody [clone: GA5]
Catalog # CSTA3670S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:GFAP
- Clonality:Monoclonal
- Clone:GA5
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:purified GFAP from porcine spinal cord
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat