Anti-Tuberin/TSC2 (Ser939) phosphate Rabbit Polyclonal Antibody
CSTA3615S
- Antibody type:Primary
- Antigen symbol:Tuberin/TSC2 (Ser939) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CSFRARSTsLNER
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Tuberin/TSC2 (Ser939) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse