Anti-APR3 Rabbit Polyclonal Antibody
Catalog # CSTA8581S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:APR3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Sequence:CSLEDPGPNFHQAHTTVIID
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: APR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human