Anti-Ephrin B1 (Tyr324/Tyr329) phosphate Rabbit Polyclonal Antibody
CSTA3481S
- Antibody type:Primary
- Antigen symbol:Ephrin B1 (Tyr324/Tyr329) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:Ephrin B2 (Tyr311/Tyr316) phosphate|Ephrin B1 (Tyr324/Tyr329) phosphate|Ephrin B3 (Tyr318/Tyr323) phosphate
- Sequence:CSGDyGHPVyIVQEM
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Ephrin B1 (Tyr324/Tyr329) phosphate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human