Anti-Cathepsin B Rabbit Monoclonal Antibody [clone: D1C7Y]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: Cathepsin B
Clonality: Monoclonal
Clone: D1C7Y
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- CSTA31718S
- CSTA31718T
- Antigen symbol:
- Cathepsin B
- Cathepsin B
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D1C7Y
- D1C7Y
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- P07858 N228-D333
- P07858 N228-D333
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together