Anti-NRF2 Rabbit Monoclonal Antibody [clone: D1Z9C]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: NRF2
Clonality: Monoclonal
Clone: D1Z9C)
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey
Specifications
- Catalog No:
- CSTA12721T
- CSTA12721S
- Antigen symbol:
- NRF2
- NRF2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D1Z9C)
- D1Z9C)
- Reactivity:
- Monkey, Human, Mouse
- Monkey, Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Sequence:
- CNSLNSDATVNTDFGDEFYSAFIAE
- CNSLNSDATVNTDFGDEFYSAFIAE
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together