Anti-SUPT16H/SPT16 Rabbit Monoclonal Antibody [clone: D7I2K]
CSTA12191S
- Antibody type:Primary
- Antigen symbol:SUPT16H/SPT16
- Clonality:Monoclonal
- Clone:D7I2K
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Sequence:CNLNRSNPKLKDLYIRPNIAQKR
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: SUPT16H/SPT16
Clonality: Monoclonal
Clone: D7I2K
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey