Order Entry
Ireland
ContactUsLinkComponent
 
Anti-SGK3 (Thr320) phosphate Rabbit Monoclonal Antibody [clone: 'D30E6]
  CSTA5642S
Anti-SGK3 (Thr320) phosphate Rabbit Monoclonal Antibody [clone: 'D30E6]
  CSTA5642S

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SGK3 (Thr320) phosphate
  • Clonality:
    Monoclonal
  • Clone:
    D30E6
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Sequence:
    KLLSLIKGVIVHRLEGVEGPSLAISDTTTtFCGT
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    Ambient
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary Antibodies
Antigen: SGK3 (Thr320) phosphate
Clonality: Monoclonal
Clone: 'D30E6
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human