Anti-MEK2 Mouse Monoclonal Antibody [clone: L38C12]
Catalog # CSTA4694S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:MEK2
- Clonality:Monoclonal
- Clone:L38C12
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:MEK1
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: MEK2
Clonality: Monoclonal
Clone: L38C12
Conjugation:
Epitope:
Hoat: Mouse
Isotype: IgG1
Reactivity: Human,Mouse,Rat,Monkey