Anti-CD105/endoglin Rabbit Monoclonal Antibody [clone: D50G1]
Catalog # CSTA4335S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:CD105/endoglin
- Clonality:Monoclonal
- Clone:D50G1
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CGLYLSPHFLQASNTIEPGQQS
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: CD105/endoglin
Clonality: Monoclonal
Clone: D50G1
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human