Anti-MMP9 Rabbit Polyclonal Antibody
CSTA3852S
- Antibody type:Primary
- Antigen symbol:MMP9
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Sequence:FQTFEGDLKWHC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human