Anti-beta-Trcp Rabbit Monoclonal Antibody [clone: D12C8]
CSTA11984S
- Antibody type:Primary
- Antigen symbol:beta-Trcp
- Clonality:Monoclonal
- Clone:D12C8
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:hbTrcp(M1 - R605)
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: beta-Trcp
Clonality: Monoclonal
Clone: D12C8
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey