Anti-TCF4/TCF7L2 Rabbit Monoclonal Antibody [clone: C9B9]
Catalog # CSTA2565S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:TCF4/TCF7L2
- Clonality:Monoclonal
- Clone:C9B9
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- ChIP:Yes
- Sequence:CRSESFRDKSRESLEEAAKRQDGG
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TCF4/TCF7L2
Clonality: Monoclonal
Clone: C9B9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human