Order Entry
Ireland
ContactUsLinkComponent
Anti-Synapsin-1 (Ser9) phosphate Rabbit Polyclonal Antibody
Anti-Synapsin-1 (Ser9) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA2311S
Supplier:  Cell Signaling
Anti-Synapsin-1 (Ser9) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA2311S
Supplier:  Cell Signaling

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Synapsin-1 (Ser9) phosphate
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Sequence:
    CYLRRRLsDSNF
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    Ambient
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Synapsin-1 (Ser9) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat