Anti-hnRNP R Rabbit Polyclonal Antibody
Catalog # CSTA5351S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:hnRNP R
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:hnRNP Q
- Sequence:CGYKSENQEFYQDTFG
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: hnRNP R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human