Anti-tubulin alpha Rabbit Monoclonal Antibody [clone: 11H10] (Alexa Fluor 555)
Catalog # CSTA5059S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:tubulin alpha
- Clonality:Monoclonal
- Clone:11H10
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Pig,Monkey,Human,Bovine,D. melanogasterm,Rat,Mouse
- Sequence:CQPDGQMPSDKTIGGGDDS
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: tubulin alpha
Clonality: Monoclonal
Clone: 11H10
Conjugation: Alexa Fluor 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey;D. melanogasterm;Zebrafish;Bovine;Pig