Anti-DRP1 (Ser637) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA4867S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:DRP1 (Ser637) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Rat
- Western blot:Yes
- Sequence:PVARKLsAREQRDC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: DRP1 (Ser637) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat