Anti-Cyclin E2 Rabbit Polyclonal Antibody
CSTA4132S
- Antibody type:Primary
- Antigen symbol:Cyclin E2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Sequence:CMTPPKSTEKPPGKH
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Cyclin E2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human