Acetone
1
Catalog # BDHB1101-1LP
Supplier: BDH US PRODUCED
Acetone
Catalog # BDHB1101-1LP
Supplier: BDH US PRODUCED
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56,2 °C (1013 hPa)
- Melting Pt:–95,4 °C
- Density:0,792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Specifications
Specifications
- Pk:1
- Cat. No.:BDHB1101-1LP
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



