tert-Butyl ethyl ether analytical standard, Supelco®
Catalog # 442795.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:102.18 g/mol
Merck Index:13,03809
Boiling Pt:71 °C (1013 hPa)
Melting Pt:–97 °C
Density:0,752 g/cm³ (20 °C)
EINECS:211-309-7
UN:1179
ADR:3,II
CAS Number:637-92-3
Formula:C₆H₁₄O
MDL Number:MFCD00009225