


Phthalaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:643-79-8
:
Formula:C₈H₆O₂
MDL Number:MFCD00003335
:134.13 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1,13 g/cm³ (20 °C)
Flash Pt:132 °C
UN:2923
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:211-402-2
:GHS06
:GHS05
:GHS09
Frequently Bought Together