N-Ethylmaleimide (NEM), Pierce™
Catalog # PIER23030
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione
:
Merck Index:13,03855
Boiling Pt:210 °C (1013 hPa)
Melting Pt:46 °C
ADR:6.1,II
:125.13 g/mol
EINECS:204-892-4
UN:2928
:GHS06
:GHS05
CAS Number:128-53-0
Formula:C₆H₇NO₂
MDL Number:MFCD00005509