1-(Trimethylsilyl)imidazole
Catalog # ROTH0012.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:140.26 g/mol
Boiling Pt:209 °C (1013 hPa)
Melting Pt:–42 °C
Density:0,95 g/cm³ (20 °C)
Flash Pt:61 °C
EINECS:242-040-3
CAS Number:18156-74-6
Formula:C₆H₁₂N₂Si
MDL Number:MFCD00005280