Order Entry
Ireland
ContactUsLinkComponent
1-(Trimethylsilyl)imidazole
1-(Trimethylsilyl)imidazole
Catalog # MOLEM50500594
Supplier:  Molekula
CAS Number:  
1-(Trimethylsilyl)imidazole
Catalog # MOLEM50500594
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :140.26 g/mol
Boiling Pt:209 °C (1013 hPa)
Melting Pt:–42 °C
Density:0,95 g/cm³ (20 °C)
Flash Pt:61 °C
EINECS:242-040-3
CAS Number:18156-74-6
Formula:C₆H₁₂N₂Si
MDL Number:MFCD00005280