Order Entry
Ireland
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)acetamide
Flammable
Corrosive
Irritant
N,O-Bis(trimethylsilyl)acetamide
Catalog # MOLE64289732-100G
Supplier:  Molekula
CAS Number:  
Flammable
Corrosive
Irritant
N,O-Bis(trimethylsilyl)acetamide
Catalog # MOLE64289732-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :203.43 g/mol
Boiling Pt:162 °C (1013 hPa)
Melting Pt:∼24 °C
Density:0,82 g/cm³ (20 °C)
Flash Pt:40 °C
EINECS:233-892-7
UN:2920
ADR:8,II
 :GHS02
 :GHS05
 :GHS07
CAS Number:10416-59-8
Formula:C₈H₂₁NOSi₂
MDL Number:MFCD00008270