
2-(4-Hydroxyphenyl)ethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:501-94-0
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00002902
:138.17 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:90…92 °C
:M-Clarity™ quality level = MQ200
EINECS:207-930-8
:GHS07
Specification Test Results
- Solubility:124 g/l (25 °C)
- Melting Point:91 - 94 °C
- Molar Mass:138.17 g/mol
Specification Test Results
Frequently Bought Together