Order Entry
Ireland
ContactUsLinkComponent
6-Propyl-2-thiouracil for synthesis, Sigma-Aldrich®
Irritant
6-Propyl-2-thiouracil for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
6-Propyl-2-thiouracil for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1.1 g/l (20 °C)
  • Melting Point:
    216 - 220 °C
  • Molar Mass:
    170.22 g/mol
  • Bulk Density:
    159 kg/m3
  • Flash Point:
    300 °C
  • pH:
    6 - 7.5 (5 g/l, H2O, 20 °C) (slurry)

Specification Test Results

Related Information

Frequently Bought Together