2-Thiouracil for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:141-90-2
:
Formula:C₄H₄N₂OS
MDL Number:MFCD00006039
:128.15 g/mol
Merck Index:12,09504
Melting Pt:340 °C
:M-Clarity™ quality level = MQ200
EINECS:205-508-8
Specification Test Results
- Solubility:7.9 g/l (25 °C)
- Melting Point:300 °C
- Molar Mass:128.14 g/mol
- Bulk Density:600 kg/m3
- Ignition Point:570 °C
Specification Test Results
Frequently Bought Together