


Furan stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-00-9
:
Formula:C₄H₄O
MDL Number:MFCD00003222
:68.08 g/mol
Merck Index:13,04317
Boiling Pt:31,4 °C (1013 hPa)
Melting Pt:–86 °C
Density:0,9371 g/cm³ (20 °C)
EINECS:203-727-3
UN:2389
ADR:3,I
:M-Clarity™ quality level = MQ200
Flash Pt:–50 °C
:GHS02
:GHS08
:GHS07
Specification Test Results
- Solubility:10 g/l (25 °C)
- Melting Point:-86 °C
- Molar Mass:68.07 g/mol
- Boiling Point:32 °C (1013 hPa)
- Vapor Pressure:670 hPa (20 °C)
- Flash Point:-36 °C
- Refractive Index:1.4207 (20 °C, 589 nm)
- Explosion Limit:2.3 - 14.3 %(V)
- Density:0.94 g/cm3 (20 °C)
- Ignition Point:390 °C
- Saturation concentration:1870 g/m3
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together