
N,N'-Diphenyl-p-phenylenediamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:74-31-7
:
Formula:C₁₈H₁₆N₂
MDL Number:MFCD00003015
:260.34 g/mol
Merck Index:12,03388
Boiling Pt:469 °C (1013 hPa)
Melting Pt:146…149 °C
Density:1,21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:200-806-4
:GHS07
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:135 °C
- Molar Mass:260.33 g/mol
- Bulk Density:560 kg/m3
- Boiling Point:220 - 225 °C (1.33 hPa)
- Density:1.3 g/cm3 (20 °C)
Specification Test Results
Related Information
Frequently Bought Together