1-(Trimethylsilyl)imidazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:18156-74-6
:
Formula:C₆H₁₂N₂Si
MDL Number:MFCD00005280
:140.26 g/mol
Boiling Pt:209 °C (1013 hPa)
Melting Pt:–42 °C
Density:0,95 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:61 °C
EINECS:242-040-3
Specification Test Results
- Solubility:(20 °C) Risk of violent reaction.
- Melting Point:-42 °C
- Molar Mass:140.25 g/mol
- Boiling Point:212 °C (decomposition)
- Density:0.95 g/cm3 (25 °C)
- Ignition Point:540 °C DIN 51794
- Viscosity Kinematic:2.5 mm2/s (25 °C)
Specification Test Results
Frequently Bought Together