2-Mercaptobenzothiazole, Sigma-Aldrich®
: Merck
:149-30-4
:
:C₇H₅NS₂
:MFCD00005781
:167.26 g/mol
:13,05893
:302 °C (1013 hPa)
:177…182 °C
:1,42 g/cm³ (20 °C)
:3077
:9,III
:M-Clarity™ quality level = MQ200
:205-736-8
:GHS07
:GHS09
:Ambient
- Solubility:0.12 g/l (25 °C)
- Melting Point:180 - 183 °C
- Molar Mass:167.25 g/mol
- Bulk Density:390 kg/m3
- Boiling Point:260 °C decomposes
- Vapor Pressure:- 0.000003 hPa (25 °C)
- Flash Point:243 °C
- Explosion Limit:15 %(V)
- Density:1.42 g/cm3 (20 °C)
- pH:7 (0.12 g/l, H2O, 25 °C)
- Ignition Point:465 °C
Frequently Bought Together